Assay Design



Gene Transcripts

Gene Symbol
WNT6
Entrez Gene ID
505075
Gene Name
Wnt family member 6
Gene Aliases
-
Chromosome Location
Chr.2: 106822096 - 106834781 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205563.1 NP_001192492.1 2 - 3 396 87

View Full Product Details