Assay Design



Gene Transcripts

Gene Symbol
TRIM68
Entrez Gene ID
538657
Gene Name
tripartite motif containing 68
Gene Aliases
-
Chromosome Location
Chr.15: 50412919 - 50427041 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205571.1 NP_001192500.1 2 - 3 545 77

View Full Product Details