Assay Design



Gene Transcripts

Gene Symbol
ERI2
Entrez Gene ID
538719
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
-
Chromosome Location
Chr.25: 18540516 - 18556531 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205577.1 NP_001192506.1 8 - 9 1026 145

View Full Product Details