Assay Design



Gene Transcripts

Gene Symbol
ETV3
Entrez Gene ID
538995
Gene Name
ETS variant transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.3: 13730452 - 13744944 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205609.1 NP_001192538.1 2 - 3 282 127

View Full Product Details