Assay Design



Gene Transcripts

Gene Symbol
LYRM1
Entrez Gene ID
508417
Gene Name
LYR motif containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 18681018 - 18731212 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205637.1 NP_001192566.1 1 - 2 165 80

View Full Product Details