Assay Design



Gene Transcripts

Gene Symbol
CYP7A1
Entrez Gene ID
510507
Gene Name
cytochrome P450, family 7, subfamily A, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.14: 24664833 - 24675169 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205677.2 NP_001192606.1 1 - 2 80 100

View Full Product Details