Assay Design



Gene Transcripts

Gene Symbol
BEST4
Entrez Gene ID
510542
Gene Name
bestrophin 4
Gene Aliases
-
Chromosome Location
Chr.3: 101224531 - 101230182 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205679.2 NP_001192608.2 6 - 7 902 80

View Full Product Details