Assay Design



Gene Transcripts

Gene Symbol
DNAJC9
Entrez Gene ID
510898
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
-
Chromosome Location
Chr.28: 29288865 - 29293350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205686.1 NP_001192615.1 1 - 2 287 67

View Full Product Details