Assay Design



Gene Transcripts

Gene Symbol
KCTD21
Entrez Gene ID
539645
Gene Name
potassium channel tetramerization domain containing 21
Gene Aliases
-
Chromosome Location
Chr.29: 17860450 - 17881538 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205693.2 NP_001192622.1 2 - 2 825 102

View Full Product Details