Assay Design



Gene Transcripts

Gene Symbol
MGAT4C
Entrez Gene ID
539661
Gene Name
MGAT4 family member C
Gene Aliases
-
Chromosome Location
Chr.5: 15662678 - 15959427 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205696.1 NP_001192625.1 2 - 3 233 105
GenBank mRNA BC112646.1 - 2 - 3 235 105

View Full Product Details