Assay Design



Gene Transcripts

Gene Symbol
RBSN
Entrez Gene ID
511484
Gene Name
rabenosyn, RAB effector
Gene Aliases
ZFYVE20
Chromosome Location
Chr.22: 57158475 - 57203348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205700.1 NP_001192629.1 6 - 7 892 86

View Full Product Details