Assay Design



Gene Transcripts

Gene Symbol
TBC1D10B
Entrez Gene ID
513858
Gene Name
TBC1 domain family member 10B
Gene Aliases
-
Chromosome Location
Chr.25: 26544203 - 26555450 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205771.1 NP_001192700.1 4 - 5 1278 73

View Full Product Details