Assay Design



Gene Transcripts

Gene Symbol
TXNDC17
Entrez Gene ID
404159
Gene Name
thioredoxin domain containing 17
Gene Aliases
TXNL5
Chromosome Location
Chr.19: 25236051 - 25238303 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205821.1 NP_001192750.1 2 - 3 302 93
GenBank mRNA AJ344102.1 - 2 - 3 176 93

View Full Product Details