Assay Design



Gene Transcripts

Gene Symbol
TRIM62
Entrez Gene ID
516886
Gene Name
tripartite motif containing 62
Gene Aliases
-
Chromosome Location
Chr.2: 120514941 - 120552835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205835.1 NP_001192764.1 1 - 2 410 64

View Full Product Details