Assay Design



Gene Transcripts

Gene Symbol
SLC25A45
Entrez Gene ID
517425
Gene Name
solute carrier family 25 member 45
Gene Aliases
-
Chromosome Location
Chr.29: 43568455 - 43575007 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205840.1 NP_001192769.1 3 - 4 435 65

View Full Product Details