Assay Design



Gene Transcripts

Gene Symbol
TRIML1
Entrez Gene ID
520202
Gene Name
tripartite motif family like 1
Gene Aliases
-
Chromosome Location
Chr.27: 17666005 - 17672948 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205868.1 NP_001192797.1 1 - 2 407 121

View Full Product Details