Assay Design



Gene Transcripts

Gene Symbol
SYNE3
Entrez Gene ID
520207
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, C21H14orf49
Chromosome Location
Chr.21: 60265774 - 60379931 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205869.2 NP_001192798.1 15 - 16 2586 66

View Full Product Details