Assay Design



Gene Transcripts

Gene Symbol
KCNS1
Entrez Gene ID
521338
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
-
Chromosome Location
Chr.13: 73470239 - 73476819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205884.2 NP_001192813.1 1 - 2 73 83

View Full Product Details