Assay Design



Gene Transcripts

Gene Symbol
AGO1
Entrez Gene ID
522972
Gene Name
argonaute RISC component 1
Gene Aliases
EIF2C1
Chromosome Location
Chr.3: 109875883 - 109914109 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205899.3 NP_001192828.1 8 - 9 1196 61
NM_001317187.2 NP_001304116.1 8 - 9 1196 61

View Full Product Details