Assay Design



Gene Transcripts

Gene Symbol
LHFPL5
Entrez Gene ID
527838
Gene Name
LHFPL tetraspan subfamily member 5
Gene Aliases
-
Chromosome Location
Chr.23: 9767592 - 9783138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205946.1 NP_001192875.1 1 - 2 418 95

View Full Product Details