Assay Design



Gene Transcripts

Gene Symbol
MFAP3L
Entrez Gene ID
529443
Gene Name
microfibril associated protein 3 like
Gene Aliases
-
Chromosome Location
Chr.8: 1827004 - 1879390 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205964.1 NP_001192893.1 1 - 2 298 68

View Full Product Details