Assay Design



Gene Transcripts

Gene Symbol
CFAP70
Entrez Gene ID
529891
Gene Name
cilia and flagella associated protein 70
Gene Aliases
TTC18
Chromosome Location
Chr.28: 29299960 - 29383242 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205969.1 NP_001192898.1 19 - 20 2458 90

View Full Product Details