Assay Design



Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
530042
Gene Name
neuroligin 1
Gene Aliases
-
Chromosome Location
Chr.1: 93145361 - 94102525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205973.2 NP_001192902.1 5 - 6 2080 105

View Full Product Details