Assay Design



Gene Transcripts

Gene Symbol
KLF3
Entrez Gene ID
530358
Gene Name
Kruppel like factor 3
Gene Aliases
-
Chromosome Location
Chr.6: 57942693 - 57978356 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205978.1 NP_001192907.1 3 - 4 727 76

View Full Product Details