Assay Design



Gene Transcripts

Gene Symbol
ZDHHC24
Entrez Gene ID
530442
Gene Name
zinc finger DHHC-type containing 24
Gene Aliases
-
Chromosome Location
Chr.29: 44573585 - 44584099 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205979.1 NP_001192908.1 1 - 2 280 54

View Full Product Details