Assay Design



Gene Transcripts

Gene Symbol
FOXRED2
Entrez Gene ID
532871
Gene Name
FAD dependent oxidoreductase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.5: 74897794 - 74919086 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206011.1 NP_001192940.1 2 - 3 799 91

View Full Product Details