Assay Design



Gene Transcripts

Gene Symbol
RBM38
Entrez Gene ID
533516
Gene Name
RNA binding motif protein 38
Gene Aliases
-
Chromosome Location
Chr.13: 58757864 - 58773500 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206023.1 NP_001192952.1 3 - 4 599 83

View Full Product Details