Assay Design



Gene Transcripts

Gene Symbol
SLC41A1
Entrez Gene ID
533907
Gene Name
solute carrier family 41 member 1
Gene Aliases
-
Chromosome Location
Chr.16: 3531277 - 3553829 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206036.1 NP_001192965.1 2 - 3 484 58

View Full Product Details