Assay Design



Gene Transcripts

Gene Symbol
RNLS
Entrez Gene ID
534778
Gene Name
renalase, FAD dependent amine oxidase
Gene Aliases
-
Chromosome Location
Chr.26: 9901178 - 10177515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206063.1 NP_001192992.1 1 - 2 174 64

View Full Product Details