Assay Design



Gene Transcripts

Gene Symbol
NOVA2
Entrez Gene ID
537783
Gene Name
NOVA alternative splicing regulator 2
Gene Aliases
-
Chromosome Location
Chr.18: 53442851 - 53469024 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206109.2 NP_001193038.2 2 - 3 230 65

View Full Product Details