Assay Design



Gene Transcripts

Gene Symbol
ATP4B
Entrez Gene ID
614308
Gene Name
ATPase H+/K+ transporting subunit beta
Gene Aliases
-
Chromosome Location
Chr.12: 86741886 - 86746925 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206140.1 NP_001193069.1 4 - 5 576 107

View Full Product Details