Assay Design



Gene Transcripts

Gene Symbol
POLR3G
Entrez Gene ID
613778
Gene Name
RNA polymerase III subunit G
Gene Aliases
-
Chromosome Location
Chr.7: 90036757 - 90081679 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206143.1 NP_001193072.1 3 - 4 298 145

View Full Product Details