Assay Design



Gene Transcripts

Gene Symbol
SMAD6
Entrez Gene ID
511279
Gene Name
SMAD family member 6
Gene Aliases
Smad6-like
Chromosome Location
Chr.10: 13544183 - 13622063 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206145.2 NP_001193074.1 1 - 2 1581 74

View Full Product Details