Assay Design



Gene Transcripts

Gene Symbol
GOPC
Entrez Gene ID
541186
Gene Name
golgi associated PDZ and coiled-coil motif containing
Gene Aliases
-
Chromosome Location
Chr.9: 33189369 - 33229409 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206157.1 NP_001193086.1 4 - 5 899 108

View Full Product Details