Assay Design



Gene Transcripts

Gene Symbol
RPRD2
Entrez Gene ID
614766
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 20203806 - 20303069 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206187.1 NP_001193116.1 2 - 3 331 141

View Full Product Details