Assay Design



Gene Transcripts

Gene Symbol
PLEKHM3
Entrez Gene ID
533312
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
PLEKHM1L
Chromosome Location
Chr.2: 96095228 - 96318519 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206217.1 NP_001193146.1 7 - 8 2555 89

View Full Product Details