Assay Design



Gene Transcripts

Gene Symbol
UCMA
Entrez Gene ID
615924
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
-
Chromosome Location
Chr.13: 27897057 - 27904337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206226.1 NP_001193155.1 2 - 3 179 66

View Full Product Details