Assay Design



Gene Transcripts

Gene Symbol
NKX6-1
Entrez Gene ID
616211
Gene Name
NK6 homeobox 1
Gene Aliases
-
Chromosome Location
Chr.6: 99307037 - 99312032 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206238.1 NP_001193167.1 1 - 2 659 95

View Full Product Details