Assay Design



Gene Transcripts

Gene Symbol
KRT36
Entrez Gene ID
520668
Gene Name
keratin 36
Gene Aliases
-
Chromosome Location
Chr.19: 41690878 - 41694867 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192525.2 NP_001179454.2 5 - 6 1028 72

View Full Product Details