Assay Design



Gene Transcripts

Gene Symbol
SRMS
Entrez Gene ID
521816
Gene Name
src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Gene Aliases
-
Chromosome Location
Chr.13: 54119642 - 54125436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192544.1 NP_001179473.1 7 - 8 1295 77

View Full Product Details