Assay Design



Gene Transcripts

Gene Symbol
HMX2
Entrez Gene ID
525631
Gene Name
H6 family homeobox 2
Gene Aliases
-
Chromosome Location
Chr.26: 42935747 - 42944278 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192615.1 NP_001179544.1 1 - 2 260 118

View Full Product Details