Assay Design



Gene Transcripts

Gene Symbol
CSF2RB
Entrez Gene ID
528530
Gene Name
colony stimulating factor 2 receptor subunit beta
Gene Aliases
-
Chromosome Location
Chr.5: 75349117 - 75372728 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192664.2 NP_001179593.2 12 - 13 1666 79

View Full Product Details