Assay Design



Gene Transcripts

Gene Symbol
KLK5
Entrez Gene ID
617418
Gene Name
kallikrein related peptidase 5
Gene Aliases
KLNC
Chromosome Location
Chr.18: 56932757 - 56942570 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206280.3 NP_001193209.2 4 - 5 725 67

View Full Product Details