Assay Design



Gene Transcripts

Gene Symbol
EPN2
Entrez Gene ID
505336
Gene Name
epsin 2
Gene Aliases
-
Chromosome Location
Chr.19: 34116025 - 34168729 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206317.2 NP_001193246.1 6 - 7 1471 106

View Full Product Details