Assay Design



Gene Transcripts

Gene Symbol
IL20RA
Entrez Gene ID
509038
Gene Name
interleukin 20 receptor subunit alpha
Gene Aliases
-
Chromosome Location
Chr.9: 74969084 - 75002224 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206342.1 NP_001193271.1 3 - 4 508 105
GenBank mRNA BC149797.1 - 3 - 4 508 105

View Full Product Details