Assay Design



Gene Transcripts

Gene Symbol
AASDHPPT
Entrez Gene ID
519816
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
-
Chromosome Location
Chr.15: 1567368 - 1594245 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206477.1 NP_001193406.1 4 - 5 760 117
GenBank mRNA DT813625.1 - 4 - 5 682 117
EV716630.1 - 2 - 3 407 117

View Full Product Details