Assay Design



Gene Transcripts

Gene Symbol
DPPA4
Entrez Gene ID
783813
Gene Name
developmental pluripotency associated 4
Gene Aliases
-
Chromosome Location
Chr.1: 54084686 - 54095829 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206481.2 NP_001193410.1 1 - 2 65 82

View Full Product Details