Assay Design



Gene Transcripts

Gene Symbol
PROX2
Entrez Gene ID
529720
Gene Name
prospero homeobox 2
Gene Aliases
-
Chromosome Location
Chr.10: 86108353 - 86139149 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192679.2 NP_001179608.2 3 - 4 1594 104

View Full Product Details