Assay Design



Gene Transcripts

Gene Symbol
QRFPR
Entrez Gene ID
529752
Gene Name
pyroglutamylated RFamide peptide receptor
Gene Aliases
GPR103
Chromosome Location
Chr.6: 4138071 - 4198240 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192681.1 NP_001179610.1 5 - 6 1253 111

View Full Product Details