Assay Design



Gene Transcripts

Gene Symbol
AWAT1
Entrez Gene ID
529893
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
DGAT2L3
Chromosome Location
Chr.X: 80203297 - 80209364 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192683.1 NP_001179612.1 6 - 7 828 104

View Full Product Details